Anti-IL3 Goat Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
BAF203
103225-034EA 837.32 USD
103225-034
Anti-IL3 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary Antibodies
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR