Anti-TNFRSF17 Goat Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
BAF193
103224-998EA 793.94 USD
103224-998
Anti-TNFRSF17 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary Antibodies
Antigen: TNFRSF17
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR