Anti-LCN2 Goat Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
BAF1857
103224-940EA 750.55 USD
103224-940
Anti-LCN2 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary Antibodies
Antigen: LCN2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR