Anti-HGFAC Goat Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
BAF1514
103224-704EA 721.63 USD
103224-704
Anti-HGFAC Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary Antibodies
Antigen: HGFAC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR