Anti-MMP2 Goat Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
BAF1488
103224-686EA 721.63 USD
103224-686
Anti-MMP2 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary Antibodies
Antigen: MMP2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR