Anti-RETN Goat Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
BAF1069
103224-284EA 779.48 USD
103224-284
Anti-RETN Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary Antibodies
Antigen: RETN
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR