Anti-IFNG Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF985
103224-182EA 663.78 USD
103224-182
Anti-IFNG Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Porcine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR