Anti-IL1RAPL1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF975-SP
103224-152EA 215.48 USD
103224-152
Anti-IL1RAPL1 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: IL1RAPL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR