Anti-TIMP4 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF974-SP
103224-148EA 215.48 USD
103224-148
Anti-TIMP4 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: TIMP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR