Anti-IFNG Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF961
103224-110EA 649.32 USD
103224-110
Anti-IFNG Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rhesus Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR