Anti-DPP4 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF954-SP
103224-084EA 244.4 USD
103224-084
Anti-DPP4 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: DPP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR