Anti-CTSB Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF953
103224-078EA 692.71 USD
103224-078
Anti-CTSB Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: CTSB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR