Anti-CTSL1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF952
103224-074EA 707.17 USD
103224-074
Anti-CTSL1 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: CTSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR