Anti-ADAM9 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF949-SP
103224-068EA 229.94 USD
103224-068
Anti-ADAM9 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: ADAM9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR