Anti-tek Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF928-SP
103223-992EA 215.48 USD
103223-992
Anti-tek Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: tek
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR