Anti-MMP14 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF918-SP
103223-972EA 215.48 USD
103223-972
Anti-MMP14 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: MMP14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR