Anti-IL12A Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF912
103223-954EA 707.17 USD
103223-954
Anti-IL12A Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: IL12A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Porcine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR