Anti-RPS6KB1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
AF8965-SP
103223-916EA 229.94 USD
103223-916
Anti-RPS6KB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: RPS6KB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR