Anti-RPS6KB1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF8964
103223-910EA 649.32 USD
103223-910
Anti-RPS6KB1 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: RPS6KB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR