Anti-MERTK Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF891-SP
103223-896EA 258.86 USD
103223-896
Anti-MERTK Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: MERTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR