Anti-BIRC5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
AF886
103223-878EA 577.02 USD
103223-878
Anti-BIRC5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR