Anti-Ccl19 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF880-SP
103223-868EA 244.4 USD
103223-868
Anti-Ccl19 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: Ccl19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR