Anti-IL10RB Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF874-SP
103223-852EA 215.48 USD
103223-852
Anti-IL10RB Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: IL10RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR