Anti-MAPK14 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
AF8691
103223-830EA 577.02 USD
103223-830
Anti-MAPK14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: MAPK14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR