Anti-PSG6 Rabbit Polyclonal Antibody

AF8598-SP
103223-802EA 140 USD
103223-802
Anti-PSG6 Rabbit Polyclonal Antibody
Type: Primary Antibodies
Antigen: PSG6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR