Anti-AXL Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF854-SP
103223-774EA 258.86 USD
103223-774
Anti-AXL Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: AXL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR