Anti-GDF5 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF853
103223-762EA 663.78 USD
103223-762
Anti-GDF5 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: GDF5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR