Anti-GIT1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
AF8508-SP
103223-732EA 215.48 USD
103223-732
Anti-GIT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: GIT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR