Anti-APP Rabbit Polyclonal Antibody

AF850
103223-728EA 409.23 USD
103223-728
Anti-APP Rabbit Polyclonal Antibody
Type: Primary Antibodies
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR