Anti-NAPSA Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
AF8489
103223-724EA 634.86 USD
103223-724
Anti-NAPSA Rabbit Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: NAPSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR