Anti-PTEN Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
AF847-SP
103223-718EA 258.86 USD
103223-718
Anti-PTEN Rabbit Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR