Anti-CCL1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF845-SP
103223-710EA 215.48 USD
103223-710
Anti-CCL1 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: CCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR