Anti-CALCA Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF8350
103223-658EA 765.02 USD
103223-658
Anti-CALCA Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR