Anti-CASP2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
AF826-SP
103223-618EA 118 USD
103223-618
Anti-CASP2 Rabbit Polyclonal Antibody
Type: Primary Antibodies
Antigen: CASP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR