Anti-XIAP Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF8221-SP
103223-582EA 244.4 USD
103223-582
Anti-XIAP Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: XIAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR