Anti-BAD Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
AF819
103223-542EA 577.02 USD
103223-542
Anti-BAD Rabbit Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: BAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR