Anti-BIRC2 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF8181
103223-534EA 707.17 USD
103223-534
Anti-BIRC2 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: BIRC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR