Anti-BIRC3 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF8171
103223-524EA 692.71 USD
103223-524
Anti-BIRC3 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: BIRC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR