Anti-NOX4 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF8158-SP
103223-508EA 215.48 USD
103223-508
Anti-NOX4 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: NOX4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR