Anti-ZMIZ1 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF8107
103223-478EA 663.78 USD
103223-478
Anti-ZMIZ1 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: ZMIZ1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR