Anti-PECAM1 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF806-SP
103223-450EA 244.4 USD
103223-450
Anti-PECAM1 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: PECAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR