Anti-MAP4K5 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF8007-SP
103223-360EA 215.48 USD
103223-360
Anti-MAP4K5 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: MAP4K5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR