Anti-MCK-2 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF798
103223-336EA 605.94 USD
103223-336
Anti-MCK-2 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR