Anti-UBQLN1 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF7969
103223-302EA 663.78 USD
103223-302
Anti-UBQLN1 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: UBQLN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR