Anti-SMAD2 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF7948
103223-288EA 663.78 USD
103223-288
Anti-SMAD2 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: SMAD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR