Anti-KLK1 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF7928
103223-250EA 663.78 USD
103223-250
Anti-KLK1 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: KLK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR