Anti-C1QTNF3 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF7925-SP
103223-240EA 215.48 USD
103223-240
Anti-C1QTNF3 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: C1QTNF3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR