Anti-IGF2 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF792
103223-236EA 707.17 USD
103223-236
Anti-IGF2 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: IGF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR