Anti-TAGLN Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF7886-SP
103223-192EA 229.94 USD
103223-192
Anti-TAGLN Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: TAGLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR