Anti-Il2rg Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF784-SP
103223-138EA 215.48 USD
103223-138
Anti-Il2rg Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: Il2rg
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR